Methyl-4'-bromobiphenyl-4-carboxylate ≥95%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Methyl 4'-bromo[1,1'-biphenyl]-4-carboxylate
, Methyl 4'-bromo-[1,1'-biphenyl]-4-carboxylate
H51920.06EA
0
GBP
H51920.06
Methyl-4'-bromobiphenyl-4-carboxylate ≥95%
Methyl 4'-bromobiphenyl-4-carboxylate
Formula:
C₁₄H₁₁BrO₂ MW: 291.14 g/mol |
MDL Number:
MFCD00496638 CAS Number: 89901-03-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...