4-Nitro-2-(trifluoromethyl)benzonitrile ≥98%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
L16318.MDEA
20.1
GBP
L16318.MD
4-Nitro-2-(trifluoromethyl)benzonitrile ≥98%
4-Nitro-2-(trifluoromethyl)benzonitrile
Formula:
C₈H₃F₃N₂O₂ MW: 216.12 g/mol |
MDL Number:
MFCD01075729 CAS Number: 320-47-8 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...