(S)-(-)-1-(1-Naphthyl)ethylamine ≥99%, ee 99%, ChiPros®
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(S)-(-)-alpha-(1-Naphthyl)ethylamine
L19301.03EA
28.6
GBP
L19301.03
(S)-(-)-1-(1-Naphthyl)ethylamine ≥99%, ee 99%, ChiPros®
(S)-(-)-1-(1-Naphthyl)ethylamine
Formula:
C₁₂H₁₃N MW: 171.24 g/mol Flash Pt: >110 °C (230 °F) |
MDL Number:
MFCD00064179 CAS Number: 10420-89-0 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...