(R)-(+)-1-(1-Naphthyl)ethylamine ≥99%, ee 98%, ChiPros®
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
(R)-(+)-α-(1-Naphthyl)ethylamine
, (R)-1-(Naphthalen-1-yl)ethanamine
L19300.06EA
101.4
GBP
L19300.06
L19300.03
(R)-(+)-1-(1-Naphthyl)ethylamine ≥99%, ee 98%, ChiPros®
(R)-(+)-1-(1-Naphthyl)ethylamine
Formula:
C₁₂H₁₃N MW: 171.24 g/mol Boiling Pt: 152…153 °C (11 mmHg) Density: 1.060 Flash Pt: >110 °C (230 °F) |
MDL Number:
MFCD00064114 CAS Number: 3886-70-2 EINECS: 223-425-5 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...