Adenine sulphate
Supplier: Molekula
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Adenine hemisulphate salt
, 6-Aminopurine hemisulphate salt
, 7H-Purin-6-amine hemisulphate
MOLE40343098-100GEA
0
GBP
MOLE40343098-100G
Adenine sulphate
Adenine sulphate
Formula:
C₅H₅N₅·½H₂SO₄ MW: 184.17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00213655 CAS Number: 321-30-2 EINECS: 206-286-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...