2-(4-Fluorophenyl)indole
Supplier: Molekula
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MOLEM34436720EA
0
GBP
MOLEM34436720
2-(4-Fluorophenyl)indole
2-(4-Fluorophenyl)indole
Formula:
C₁₄H₁₀FN |
MDL Number:
MFCD00068178 CAS Number: 782-17-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...