Diethyl succinate, Msynth® plus, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ400
8.45009.0250EA
36.1
GBP
8.45009.0250
8.45009.1000
Diethyl succinate, Msynth® plus, Sigma-Aldrich®
Diethyl succinate
Formula:
C₈H₁₄O₄ MW: 174.2 g/mol Boiling Pt: 217 °C (1013 hPa) Melting Pt: –19 °C Density: 1.04 g/cm³ (20 °C) Flash Pt: 80 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009208 CAS Number: 123-25-1 EINECS: 204-612-0 Merck Index: 12,09038 |
Specification Test Results
Solubility | 2.00 g/l (25 °C) |
Melting Point | -20 °C |
Molar Mass | 174.19 g/mol |
Boiling Point | 215 - 217 °C (1013 hPa) |
Vapor Pressure | 1.33 hPa (55 °C) |
Flash Point | 80 °C |
Refractive Index | 1.42 (20 °C, 589 nm) |
Density | 1.04 g/cm3 (20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...