ALFAB21509

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

B21509.14EA 70.1 GBP
B21509.14 B21509.06
ALFAB21509
Purity: 98%
CAS No.: 452-10-8
Molecular Formula: C7H6F2O
Formula Weight: 144.12
Flash Point: 51°(123°F)
Refractive Index: 1.4700
Boiling Point: 53°/18mm
Beilstein Reference: 2518264
UN No.: 1993
MDL No.: MFCD00010731
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR