ALFAB23750

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

B23750.06EA 86.6 GBP
B23750.06
ALFAB23750
Purity: 98+%
CAS No.: 437-82-1
Molecular Formula: C7H6F2O
Formula Weight: 144.12
Flash Point: 42°(108°F)
Boiling Point: 70-72°/56mm
UN No.: 1993
MDL No.: MFCD00142846
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR