ALFAB22916

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

B22916.03EA 27.6 GBP
B22916.03 B22916.06
ALFAB22916
Purity: 97+%
CAS No.: 134364-69-5
Molecular Formula: C7H6F2O
Formula Weight: 144.12
Flash Point: 62°(144°F)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR