1-Fluoro-2-nitrobenzene for synthesis

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: o-Fluoronitrobenzene2-Fluoronitrobenzene

M-Clarity™ quality level = MQ200

8.21798.9999
1-Fluoro-2-nitrobenzene for synthesis
1-Fluoro-2-nitrobenzene
Formula: C₆H₄FNO₂
MW: 141.1 g/mol
Boiling Pt: 212 °C (1013 hPa)
Melting Pt: –8 °C
Density: 1.33 g/cm³ (20 °C)
Flash Pt: 94 °C
Storage Temperature: Ambient
MDL Number: MFCD00007048
CAS Number: 1493-27-2
EINECS: 216-088-0
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Solubility (20 °C) slightly soluble
Melting Point -8 °C
Molar Mass 141.1 g/mol
Boiling Point 215 °C (1013 hPa)
Flash Point 94 °C
Refractive Index 1.5323 (17 °C, 589 nm)
Density 1.33 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR