2,6-Dichloroaniline
Supplier: EHRENSTORFER GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
EHERC12322600EA
85
GBP
EHERC12322600
2,6-Dichloroaniline
2,6-Dichloroaniline
Formula:
C₆H₅Cl₂N MW: 162.02 g/mol Boiling Pt: 220 °C (1013 hPa) Melting Pt: 35…38 °C Density: 1.526 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007675 CAS Number: 608-31-1 EINECS: 210-160-5 UN: 3442 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...