Autoclave, benchtop, Labclave 23
Supplier: LTE Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
481-0633EA 0 GBP
LTEALAB/23/01
Autoclave, benchtop, Labclave 23
The Labclave 23 is a low-cost 23 litre laboratory autoclave from LTE Scientific. It has been designed to offer effective and fast sterilisation of many laboratory load types including fluids, discard, glassware and instruments. Taking up a floor area of just 460×395 mm, Labclave 23 has to be one of the most compact and versatile units available.
  • Complete record of every cycle is produced on the built in thermal printer
  • Cooled condensate is discharged directly to drain if required, although a waste container is provided as standard
  • The ingenious door closure system seals and locks the door at the push of a button
Four types of cooling method are available on the Labclave 23:
Slow cooling, no vacuum; exhaust valve is permanently closed during cooling
Slow cooling, vacuum; exhaust valve opens at atmospheric pressure at which point the vacuum pump initiates
Fast cooling, no vacuum; exhaust valve is open during cooling
Fast cooling, vacuum; exhaust valve is open and vacuum pump is on during cooling
Flexible control system: Labclave 23 comes with 8 preset programs covering the most popular sterilisation cycles. In addition, a ‘free cycle’ is also provided, allowing the user to set their own specific cycle parameters quickly and easily.
Safety: Labclave 23 is fitted with pressure, temperature and timed door safety systems, ensuring that the door cannot be opened until the chamber has reached atmospheric pressure and 90 °C. A low water protection device is also fitted.