1,3-Dichloropropene, Supelco®

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger


M-Clarity™ quality level = MQ100

48530-UEA 85.3 GBP
48530-U
1,3-Dichloropropene, Supelco®
1,3-Dichloropropene
Formula: C₃H₄Cl₂
MW: 110.97 g/mol
Boiling Pt: 108 °C (1013 hPa)
Melting Pt: –83.9 °C
Density: 1.25 g/cm³ (0 °C)
Storage Temperature: Ambient
CAS Number: 542-75-6
EINECS: 208-826-5
UN: 2047
ADR: 3,II
Merck Index: 13,03101

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR