Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Burette stopcock, straight or lateral

Supplier: Brand
BISTABIL
Borosilicate glass 3.3, clear

  • Interchangeable key
  • With glass key
  • Key with retention device

Stopcock housing with precision burette tip and key.



 

ORDER

Expand All / Collapse All

Straight stopcocks
For Pk VWR Catalog Number Quantity
burettes of 2 - 10 ml capacity 1 BRND81815
burettes of 25 ml capacity 1 BRND81817
burettes of 50 ml capacity 1 BRND81819

Lateral stopcocks
For Pk VWR Catalog Number Quantity
burettes of 2 - 10 ml capacity 1 BRND81805
burettes of 25 ml capacity 1 BRND81807
burettes of 50 ml capacity 1 BRND81809

Spare Keys
Description Spigot For Pk VWR Catalog Number Quantity
Spare glass key for straight stopcocks, for burettes of 2-10 ml, 25 ml and 50 ml capacity 3NS/12 NS for burettes of capacity 2-10 ml, 25 ml and 50 ml 1 BRND81053
Spare glass key for lateral stopcocks, for burettes of 2-10 ml capacity 3NS/12 NS for burettes of capacity 2-10 ml 1 BRND81140
Spare glass key for lateral stopcocks, for burettes of 25 ml capacity, 3NS/12 NS - for burettes of capacity 25 ml 1 BRND81141
Spare glass key for lateral stopcocks, for burettes of 50 ml capacity 3NS/12 NS for burettes of capacity 50 ml 1 BRND81142
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping