1-Bromo-3-chloro-2-methylpropane for synthesis
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
M-Clarity™ quality level = MQ200
8.02622.0250EA
137
GBP
8.02622.0250
8.02622.9999
1-Bromo-3-chloro-2-methylpropane for synthesis
1-Bromo-3-chloro-2-methylpropane
Formula:
C₄H₈BrCl MW: 171.46 g/mol Boiling Pt: 157…159 °C (1013 hPa) Density: 1.4839 g/cm³ (20 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000955 CAS Number: 6974-77-2 EINECS: 230-224-6 |
Specification Test Results
Solubility | (20 °C) insoluble |
Molar Mass | 171.46 g/mol |
Boiling Point | 157 - 159 °C (1013 hPa) |
Flash Point |
|
Refractive Index | 1.4845 (20 °C, 589 nm) |
Density | 1.48 g/cm3 (20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...