n-Undecane, EMSURE® for analysis, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Undecane

M-Clarity™ quality level = MQ300

1.09795.0100EA 155 GBP
1.09795.0100
n-Undecane, EMSURE® for analysis, Supelco®
n-Undecane
Formula: H₃C(CH₂)₉CH₃
MW: 156.31 g/mol
Boiling Pt: 196 °C (1013 hPa)
Melting Pt: –26.7 °C
Density: 0.7416 g/cm³ (20 °C)
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00008959
CAS Number: 1120-21-4
EINECS: 214-300-6
UN: 2330
ADR: 3,

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Melting Point -26 °C
Molar Mass 156.31 g/mol
Boiling Point 195 °C (1013 hPa)
Vapor Pressure 0.6 hPa (20 °C)
Flash Point 60 °C
Explosion Limit 0.6 - 6.5 %(V)
Density 0.74 g/cm3 (20 °C)
Ignition Point 220 °C
Viscosity kinematic  - 7 mm2/s (40 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR