Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aspirator bottles with screw cap and spigot

Supplier: Brand
HDPE, translucent with blue screw cap and tightly fitting spigot.

  • Good chemical resistance
  • Heat resistant up to +80 °C (+120 °C for short periods)
  • Heavy walled, very tough, a deeper closure with insert seal increasing leakproof performance

Ideal for storing and dispensing liquids. Also recommended for use as settling bottles. Equipped with a carrying handle, the 25 and 50 litre sizes have two carrying handles.




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Capacity Height Ø ext. Neck I-Ø Pk VWR Catalog Number Quantity
10000 ml 420 mm 210 mm 54 mm 1 BRND131174
5000 ml 335 mm 165 mm 45 mm 1 BRND131170
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty