n-Hexane reference substance for gas chromatography, Supelco®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
M-Clarity™ quality level = MQ100
1.09687.0005EA
221
GBP
1.09687.0005
n-Hexane reference substance for gas chromatography, Supelco®
n-Hexane
Formula:
H₃C(CH₂)₄CH₃ MW: 86.18 g/mol Boiling Pt: 69 °C (1013 hPa) Melting Pt: –94.3 °C Density: 0.659 g/cm³ (20 °C) Flash Pt: –22 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009520 CAS Number: 110-54-3 EINECS: 203-777-6 UN: 1208 ADR: 3,II Merck Index: 13,04712 |
![]() |
Specification Test Results
Solubility | 0.0095 g/l (20 °C) |
Melting Point | -94.3 °C |
Molar Mass | 86.18 g/mol |
Boiling Point | 69 °C (1013 hPa) |
Vapor Pressure | 160 hPa (20 °C) |
Flash Point | -22 °C |
Refractive Index | 1.375 |
Explosion Limit | 1.0 - 8.1 %(V) |
Density | 0.66 g/cm3 (20 °C) |
pH | (H2O) not applicable |
Ignition Point | 240 °C |
Viscosity kinematic | 0.50 mm2/s (20 °C) |
Hazard Class | 3 |
Hazard Class Description | Flammable liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...