n-Hexane, Prepsolv® for preparative chromatography, Supelco®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
M-Clarity™ quality level = MQ200
1.04394.9030EA
1610
GBP
1.04394.9030
n-Hexane, Prepsolv® for preparative chromatography, Supelco®
n-Hexane
Formula:
H₃C(CH₂)₄CH₃ MW: 86.18 g/mol Boiling Pt: 69 °C (1013 hPa) Melting Pt: –94.3 °C Density: 0.659 g/cm³ (20 °C) Flash Pt: –22 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009520 CAS Number: 110-54-3 EINECS: 203-777-6 UN: 1208 ADR: 3,II Merck Index: 13,04712 |
![]() |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...