n-Heptane, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger


M-Clarity™ quality level = MQ200

SIAL246654-100MLEA 52.3 GBP
SIAL246654-100ML SIAL246654-1L SIAL246654-2L
n-Heptane, Sigma-Aldrich®
n-Heptane
Formula: H₃C(CH₂)₅CH₃
MW: 100.2 g/mol
Boiling Pt: 98.4 °C (1013 hPa)
Melting Pt: –90.6 °C
Density: 0.6838 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009544
CAS Number: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3,II
Merck Index: 13,04679
https://uk.vwr.com/stibo/web/std.lang.all/64/15/736415.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR