Dichloromethane for pesticide residue analysis
Supplier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Methylene chloride
152646CEA
0
GBP
152646C
Dichloromethane for pesticide residue analysis
Dichloromethane
Formula:
CH₂Cl₂ MW: 84.93 g/mol Boiling Pt: 39.8 °C (1013 hPa) Melting Pt: –95 °C Density: 1.322 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000881 CAS Number: 75-09-2 EINECS: 200-838-9 UN: 1593 ADR: 6.1,III Merck Index: 12,06140 |
![]() |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...