Formic acid
Supplier: EHRENSTORFER GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Methanoic acid
EHERC13913000EA
99.1
GBP
EHERC13913000
Formic acid
Formic acid
Formula:
HCO₂H MW: 46.03 g/mol Boiling Pt: 100…101 °C (1013 hPa) Melting Pt: 8.2…8.4 °C Density: 1.22 g/cm³ (25 °C) Flash Pt: 48 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003297 CAS Number: 64-18-6 EINECS: 200-579-1 UN: 1779 ADR: 8,II REACH: 01-2119491174-37 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...