Hexyl acetate for synthesis, Sigma-Aldrich®

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Acetic acid hexyl ester

M-Clarity™ quality level = MQ200

8.20555.9999
Hexyl acetate for synthesis, Sigma-Aldrich®
Hexyl acetate
Formula: C₈H₁₆O₂
MW: 144.21 g/mol
Boiling Pt: 338 °C (1013 hPa)
Melting Pt: –80 °C
Density: 0.87 g/cm³ (20 °C)
Flash Pt: 56 °C
Storage Temperature: Ambient
MDL Number: MFCD00009524
CAS Number: 142-92-7
EINECS: 205-572-7
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Solubility 0.4 g/l (20 °C)
Melting Point -80 °C
Molar Mass 144.21 g/mol
Boiling Point 171.5 °C (1013 hPa)
Vapor Pressure 5 hPa (20 °C)
Flash Point 56 °C
Refractive Index 1.409 (20 °C, 589 nm)
Density 0.87 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR