Acetylacetone ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
2,4-Pentanedione
, ACAC
20092.230EA
133
GBP
20092.230
Acetylacetone ≥99.0%, AnalaR NORMAPUR® analytical reagent
Acetylacetone
Formula:
CH₃COCH₂COCH₃ MW: 100.12 g/mol Boiling Pt: 140.4 °C (1013 hPa) Melting Pt: –23 °C Density: 0.96945 g/cm³ (20 °C) Flash Pt: 30 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008787 CAS Number: 123-54-6 EINECS: 204-634-0 UN: 2310 ADR: 3 (6.1),III Merck Index: 13,00083 |
Specification Test Results
Assay (on anhydrous substance) | Min. 99.0 % |
Evaporation residue | Max. 50 ppm |
Water | Max. 0.2 % |
Cu (Copper) | Max. 1 ppm |
Fe (Iron) | Max. 1 ppm |
Pb (Lead) | Max. 1 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...