Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetone ≥99.8%, ARISTAR® for trace analysis

Supplier: VWR Chemicals
Danger
Synonyms: 2-PropanoneDMKDimethyl ketonePropanone



Formula: CH₃COCH₃
MW: 58.08 g/mol
Boiling Pt: 56,2 °C (1013 hPa)
Melting Pt: -95,4 °C
Density: 0,792 g/cm³ (20 °C)
Flash Pt: < -20 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://uk.vwr.com/stibo/web/std.lang.all/06/07/720607.jpg
Specification Test Results
Assay Min. 99.8 %
Identity (IR) Conforms
Resistivity (MOhm-cm) Min. 5.0
Colour Max. 10 APHA
Water Max. 0.10 %
Free acid Min. 20 ppm
Free alkali Min. 2 ppm
Aldehydes (as HCHO) Max. 10 ppm
Evaporation residue Max. 5 ppm
Cl (Chloride) Max. 0.5 ppm
SO4 (Sulphate) Max. 1 ppm
Heavy metals (as Pb) Max. 0.1 ppm
Ethanol Max. 100 ppm
Methanol Max. 500 ppm
Ag (Silver) Max. 0.01 ppm
Al (Aluminium) Max. 0.05 ppm
As (Arsenic) Max. 0.01 ppm
Au (Gold) Max. 0.02 ppm
B (Boron) Max. 0.01 ppm
Ba (Barium) Max. 0.005 ppm
Be (Beryllium) Max. 0.01 ppm
Bi (Bismuth) Max. 0.02 ppm
Ca (Calcium) Max. 0.05 ppm
Cd (Cadmium) Max. 0.005 ppm
Co (Cobalt) Max. 0.005 ppm
Cr (Chromium) Max. 0.005 ppm
Cu (Copper) Max. 0.005 ppm
Fe (Iron) Max. 0.02 ppm
Ga (Gallium) Max. 0.01 ppm
In (Indium) Max. 0.01 ppm
K (Potassium) Max. 0.02 ppm
Li (Lithium) Max. 0.01 ppm
Mg (Magnesium) Max. 0.005 ppm
Mn (Manganese) Max. 0.005 ppm
Mo (Molybdenum) Max. 0.005 ppm
Na (Sodium) Max. 0.05 ppm
Ni (Nickel) Max. 0.005 ppm
Pb (Lead) Max. 0.005 ppm
Pt (Platinum) Max. 0.0500 ppm
Sb (Antimony) Max. 0.01 ppm
Sn (Tin) Max. 0.02 ppm
Sr (Strontium) Max. 0.005 ppm
Ti (Titanium) Max. 0.02 ppm
Tl (Thallium) Max. 0.01 ppm
V (Vanadium) Max. 0.01 ppm
Zn (Zinc) Max. 0.01 ppm
Zr (Zirconium) Max. 0.02 ppm
Substances reducing KMnO4 (as O) Max. 2.50 ppm

Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle SAFEBREAK 2,5 l 451004L
Get a discount if you order 4 or more. Login in now for details.
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty