Nonyl acetate
Supplier: Roth Carl
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Acetic acid nonyl ester
ROTH9434.1EA
0
GBP
ROTH9434.1
Nonyl acetate
Nonyl acetate
Formula:
C₁₁H₂₂O₂ MW: 186.29 g/mol Boiling Pt: 208…212 °C (1013 hPa) Melting Pt: –26 °C Density: 0.868 g/cm³ (20 °C) |
MDL Number:
MFCD00027340 CAS Number: 143-13-5 EINECS: 205-585-8 Merck Index: 12,06773 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...