Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Phenylpropyl acetate

Supplier: Merck
Synonyms: 3-Phenyl-1-propyl acetate



Formula: C₁₁H₁₄O₂
MW: 178,23 g/mol
Boiling Pt: 252,3 °C (1013 hPa)
Melting Pt: –60 °C
Density: 1,025 g/cm³ (16 °C)
MDL Number: MFCD00026216
CAS Number: 122-72-5
EINECS: 204-569-8
Specification Test Results
Solubility 0.69 g/l (20 °C)
Molar Mass 178.23 g/mol
Boiling Point 245 °C
Vapor Pressure < - 0.01 hPa (20 °C)
Flash Point 130 °C
Density 1.01 g/cm3 (20 °C)
Ignition Point 445 °C
Synonyms Hydroxycinnamyl acetate, ß-Phenylpropyl acetate
Hill Formula C11H14O2

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle 100 ml 8.14440.0100
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty