Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,2,2-Trifluoroethyl acetate ≥97%

Supplier: Alfa Aesar
Danger
Synonyms: Acetic acid (2,2,2-trifluoroethyl) ester



Formula: C₄H₅F₃O₂
MW: 142,08 g/mol
Boiling Pt: 78 °C
Density: 1,258
Flash Pt: 11 °C (51 °F)
Storage Temperature: Ambient
MDL Number: MFCD00198018
CAS Number: 406-95-1
UN: 3272
ADR: 3,II

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 100 g L16871.22
Glass bottle for solids 25 g L16871.14
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Formula weight 142.08
CAS No. 406-95-1
Purity 97%
Refractive index 1.3200
Flash point 11°(51°F)
Boiling point 78°
Molecular formula C4H5F3O2
MDL No. MFCD00198018
UN No. 3272
Beilstein reference 1757502

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty