Phenylmercury acetate

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger


M-Clarity™ quality level = MQ200

8.07588.0050EA 100 GBP
8.07588.0050 8.07588.9999
Phenylmercury acetate
Phenylmercury acetate
Formula: C₈H₈HgO₂
MW: 336.74 g/mol
Melting Pt: 146…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00008691
CAS Number: 62-38-4
EINECS: 200-532-5
UN: 1674
ADR: 6.1,II

Order Now

Specification Test Results

Solubility 2 g/l (20 °C)
Melting Point 146 - 150 °C
Molar Mass 336.74 g/mol
Bulk Density 800 kg/m3
Vapor Pressure 1.2 kPa (35 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR