5-Fluoro-2-nitrobenzoic acid
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
COBBAN-1062-100GEA
0
GBP
COBBAN-1062-100G
COBBAN-1062-5G
COBBAN-1062-25G
5-Fluoro-2-nitrobenzoic acid
5-Fluoro-2-nitrobenzoic acid
Formula:
C₇H₄FNO₄ MW: 185.11 g/mol |
MDL Number:
MFCD00055635 CAS Number: 320-98-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...