Isobutanol ≥99.5%

Supplier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 2-Methyl-1-propanoliso-Butanoliso-Butyl alcohol

538132-1LEA 0 GBP
538132-1L
Isobutanol ≥99.5%
Isobutanol
Formula: (CH₃)₂CHCH₂OH
MW: 74.12 g/mol
Boiling Pt: 107.9 °C (1013 hPa)
Melting Pt: –108 °C
Density: 0.802 g/cm³ (20 °C)
Flash Pt: 28 °C
Storage Temperature: Ambient
MDL Number: MFCD00004740
CAS Number: 78-83-1
EINECS: 201-148-0
UN: 1212
ADR: 3,III
Merck Index: 13,05148
https://uk.vwr.com/stibo/web/std.lang.all/12/85/2301285.jpg

Order Now

Specification Test Results

Autoignition Temperature 801 °F
Identity (IR) complying
Assay (GC) Min. 99.50 %
Density (D 20/4) 0.802 - 0.804
Non-volatile matter Max. 0.05 g/l
Water (Karl Fischer) Max. 0.05 %
Free acid (as CH₃COOH) Max. 0.003 %
1-Butanol (GC) Max. 0.30 %
Carbonyl compounds (as Butyraldehyde) Max. 0.07 %
APHA Max. 5

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR