Sulphuric acid ≥90-91%, J.T.Baker®
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
6147.2500PLEA
0
GBP
6147.2500PL
Sulphuric acid ≥90-91%, J.T.Baker®
Sulphuric acid
Formula:
H₂SO₄ MW: 98.08 g/mol Density: 1.82 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064589 CAS Number: 7664-93-9 EINECS: 231-639-5 UN: 1830 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...