Would you like to log in again?
Drum or carboy funnels
Supplier: Brand
PP or HDPE, translucent.
  • Very good chemical resistance (PP)
  • Good chemical resistance (HDPE)
  • Quick and easy to clean
For filling containers with large quantities of liquid or powdered substances quickly.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Funnel O-Ø Material Stem O-Ø Stem length Pk VWR Catalog Number Quantity
400 mm PP 40 mm 150 mm 1 BRND147055
440 mm HDPE 35 mm 140 mm 1 BRND147060
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
WRITE A REVIEW
Need help?
  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.
VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.
Please confirm the industry you work in
Example: Pharmaceutical
Please confirm your role
Example: Clinical Trial Assistant
Please confirm your location
Example: New York, NY
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty