Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Safety wash bottles, wide neck, printed

Supplier: VWR Collection
VWR®
LDPE, translucent, with PP vented screw cap.

LDPE, translucent, with PP vented spray cap

  • Chemical name in English, French, German and Spanish languages
  • Chemical formula, CAS number
  • Hazard pictogram, signal word, hazard statements H; precaution statements P

With safety imprint in accordance with regulation (EC) No. 1272/2008 (GHS). Dispensing tube with jet tip made of PP to optimise backflow of the medium that can be cut to length.

  • Chemical name in English, French, German and Spanish languages
  • Chemical formula, CAS number
  • GHS complaint symbols




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Imprint Capacity Colour code Thread Pk VWR Catalog Number Quantity
Acetone 250 ml Red GL 45 5 215-2535
Acetone 500 ml Red GL 45 5 215-2536
Distilled water 250 ml Natural GL 45 5 215-2537
Distilled water 500 ml Natural GL 45 5 215-2538
Ethanol 250 ml Orange GL 45 5 215-2539
Ethanol 500 ml Orange GL 45 5 215-2540
Isopropanol 500 ml Blue GL 45 5 215-2548
Methanol 500 ml Green GL 45 5 215-2546
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty