Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pocket torches, Mag-Lite®

Mag-Lite® Mag-Lite® Micro-Mag Mag-Lite® Solitaire
This product range provides you with the right light for any application. The computer-designed reflectors provide consistent luminous power and allow stepless switching between floodlight and spot-light in both the halogen and the krypton bulb designs.

  • High-strength
  • Corrosion-resistant aluminium
  • Switch with self-cleaning contacts (D cell)
  • O-rings for protection against splashing water
  • Spare bulb in the attached cap

Delivery information: Supplied with batteries.

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Description Height Pk VWR Catalog Number Quantity
Pocket torch 80 mm 1 139-2100
Pocket torch 125 mm 1 139-2101
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web. will publish your name as user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY


Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty