Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Amino acid analyser, L-8900

Supplier: VWR Collection
The L-8900 is a high-speed amino acid analyser with high sensitivity that is also capable of analysing physiological fluids and protein hydrolysates. It uses a reaction column filled with beads as opposed to a reaction coil. This provides a high surface area for reagent mixing resulting in a homogeneous flow which eliminates peak broadening that can occur with a coil. The ion exchange resin in the column is based on 3 µm particles that guarantee excellent resolution of the sample peaks enhancing the quality of the analysis. An internal shut off procedure acts independently of the control software to prevent column contamination.

  • High sensitivity, high speed analysis; amino acid determinations in 30 min, physiological fluid in 110 min
  • Diamond packed column without tendency to block, long column life reducing costs
  • Injection volumes from 0,1 to 100 µl, 200 place autosampler

The instrument incorporates a variable volume auto sampler, Peltier column oven and a two-channel detector with a reference wavelength for long lamp life. All necessary buffers and reagents are degassed before use and are kept under an inert gas atmosphere. Consumables, columns and the liquid waste containers can be replaced via the front door so no additional space is required at the side for maintenance. A unique post column delivery design mixes the two ninhydrin reagents just prior to the amino acid derivatisation thereby extending reagent storage by up to one year without the need for refrigeration.

Not available through VWR Int. in Denmark

Ordering information: Different types of columns are available on request. The delivery includes one 1 litre PE bottle for buffers B1-B4, B6 (empty). Hitachi AAA driver for OpenLAB CDS - factory production test for shipment to be conducted with PH analytical method.




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER


The main product you are looking for is no longer available for purchase. Accompanying products and services may be found elsewhere on the page.

ACCESSORIES

Expand All / Collapse All

Accessories
Description Pk VWR Catalog Number Quantity
Analogue output board 1 560-0180
Brown glass reagent bottle 15 560-0183
Cooling unit for AAA autosampler (after sales mounting) 1 903-0100 Unavailable
Plastic buffer bottle, 1 l 1 560-0184
TDE2 coil reactor 1 560-0182
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty