You Searched For: Syringe Dilutors
Syringe dilutors are ideal when delivering precise amounts in smaller sample preparation containers. Using a positive displacement system and plunger-style tip, the devices perform a simple single-step operation. Available in different volumes, the liquid handling instruments have inert fluid paths that minimize interior dilution residue that could contaminate future tests. With an advanced control system, the syringe dilutors program specific volumes desired for high accuracy and reproducibility in the laboratory.
Syringe dilutors are ideal when delivering precise amounts in smaller sample preparation containers. Using a positive displacement system and plunger-style tip, the devices perform a simple single-step operation. Available in different volumes, the liquid handling instruments have inert fluid paths that minimize interior dilution residue that could contaminate future tests. With an advanced control system, the syringe dilutors program specific volumes desired for high accuracy and reproducibility in the laboratory.
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Supplier: HAMILTON BONADUZ
Description: The Microlab® 600 dual syringe diluter is a semi-automated liquid handling instrument capable of dilutions up to 1:50000 in a single step. The precisi...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on 0800 22 33 44.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on 0800 22 33 44
Additional Documentation may be needed to purchase this item. A VWR representative will contact you if needed.
Additional Documentation may be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
Product(s) marked with this symbol are discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalogue Number listed above. If you need further assistance, please call VWR Customer Service on 0800 22 33 44.