Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Block heaters, analogue, SBH130

Supplier: Stuart
Analogue block heaters for precise heating of test tubes, microcentrifuge tubes, cuvettes and microtitre plates, units provide uniform and stable temperatures.

  • Hot warning light will flash whenever the temperature of the blocks is above 50 °C
  • BioCote® antimicrobial protection
  • Temperature stability and uniformity at 37 °C is ±0,1 °C within the block and across different blocks

IP Protection Class according to DIN EN 60529: IP 20

Ordering information: Supplied with block extraction tool. Blocks must be ordered separately.

*RT = Ambient




 

ORDER

Expand All / Collapse All
Type No. of blocks Temp. range Temp. stability Heating capacity W×D×H Weight Pk VWR Catalog Number Quantity
SBH130 2 RT* +8...130 °C ±0,1 °C 0,3 kW 235×280×115 mm 2,1 kg 1 460-0003
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty