Determination of anaerobic degradation processes, OxiTop® control AN 6/AN 12
Supplier: WTW
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OxiTop®
705-1008EA 15300 GBP
WTWA208227 WTWA208225
Determination of anaerobic degradation processes, OxiTop® control AN 6/AN 12
Measurement of aneaerobic degradation, OxiTop® AN6, AN12, and biogas determination. Anaerobic degradation processes take place in the absence of oxygen. In order to be able to fill the gas space above the sample with inert gas the measuring bottle has side-mounted connections, which are sealed with septa. When anaerobic degradation has taken place the dissolved CO₂ can be driven off and then removed from the gas space by means of a CO₂ absorber. The resulting pressure difference is proportional to the CO₂ concentration, the remaining over-pressure is proportional to the methane concentration. The degradation process can be comfortably followed in the “pressure” operating mode.
  • Controller has a range of between 500 - 1350 hPa
  • The head flashes for 5 seconds when accessed by controller
Delivery information: The AN sets include the ready to use package of sample bottles and adapters, stirrer platform and stirrers, OxiTop® Control heads, OC110 controller, Achat OC software, with appropriate accessories (thermostatic cabinet not included).