VWR® ND13, Screw Neck Vials
Supplier: VWR
Ratings:
Avg. Ratings: 5.0 (1 Review)
Total Ratings: 1
Avg. Ratings: 5.0 out of 5
Was this review helpful? Yes No
Yes: 1 No: 0 
Yes, I recommend
Parfait en stockage ou comme vial de rinçage en GC
VWR®
548-0051EA 0 GBP
548-0051 548-0521
VWR® ND13, Screw Neck Vials
Vials
These vials are intended for use on instruments from the following manufacturers: Dionex, Shimadzu, Spark, Varian, VWR®/Hitachi, Waters® (Wisp 48-position carousel), more.
  • Any combination of 4 ml screw neck vial ND13, with one of the 13 mm PP screw seals, can be ordered as a 2 in 1 kit
  • Barcode labelled vials can also be supplied on request
  • For storage purposes, also available with closed top screw seals
Delivery information: Kits include: 4 ml glass screw cap vial (14,7×45 mm), PP black screw cap with 8,5 mm central hole or closed top and septum.