Magnifier for glass thermometers
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
620-0777EA 32.1 GBP
VWRI620-0777
Magnifier for glass thermometers
Simple and solid stainless steel magnifier for easy reading of thermometers with fractional degrees.
  • Eliminates distortion and reading errors
  • Markings are three times larger than actual size
  • Mounts on to any thermometer with diameter from 6 to 7 mm
Suitable for any application, where fine divisions are read and accuracy counts.