Surgical masks, PP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
114-3439EA 0 GBP
ZVGA30961-00
Surgical masks, PP
Made from non-woven, spunbound PP, they are comfortable to wear, safe and highly efficient.
  • Practical dispenser
  • Adjustable nose bar suitable for spectacle wearer
  • Bacterial filtration efficiency > 98 %
Disposable three ply surgical masks particularly suitable for use in the health, high-tech and food industries.
Certifications: EN 14683: 2005