Nitrile gloves, Showa N-Dex® 7705PFT
Supplier: Showa
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
112-3256EA 47.7 GBP
BESTI7705PFTXL-10 BESTI7705PFTM-08
Nitrile gloves, Showa N-Dex® 7705PFT
Free from the accelerators common in all disposable nitrile gloves, which have been linked to workplace conditions like Type IV dermatitis. Also free from natural rubber latex therefore preventing Type I allergic reactions linked to latex proteins. The accelerator-free formulation creates a luminous, high visibility green glove.
  • Non sterile, powder-free, textured fingertips
  • Greater grip, comfort and dexterity
  • Low modulus formulation provides a comfortable fit
  • Ambidextrous
Certifications: EN 374; Cat. II; AQL 1,5