Gel storage systems, BioDesignGelFile™
Supplier: BioDesign
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BioDesignGelFile™
BDESG303EA 120 GBP
BDESG303 BDESG301
Gel storage systems, BioDesignGelFile™
Binders and Indexes
The BioDesignGelFile™ binder is a padded notebook containing 20 vinyl gel protector sheets.
  • Ideal for storing, organising and displaying dried gels
  • Clear label holder on the spine allows for personalisation of the binder
  • Chemically inert gel protector sheets have firm transparent pockets, a white background and ultra smooth seams and edges
The gels are held flat for long-term storage without cracking or curling and are protected from moisture, accidental spills and possible damage.