Saran™ wrap
Supplier: SABIC Snij-Unie
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Saran Saran™
129-3850
Saran™ wrap
Seals General Purpose Sealing Films
This is a total barrier film that has been specially developed for use in gel drying applications.
  • Less clingy than other films, allowing it to be easily removed from gel surfaces without causing the gel to crack