You Searched For: Tangential Flow Cassettes

Stackable tangential flow cassettes with sifting membranes are ideal for operations that require separating or purifying biomolecules. The interior cellulose and plastic based membranes are compatible with a wide range of chemical agents and remain stable within a broad pH range. Used in a wide range of biological fields, the tangential flow cassettes guarantee high flow rates, low hold-up volumes, and low protein binding.

Stackable tangential flow cassettes with sifting membranes are ideal for operations that require separating or purifying biomolecules. The interior cellulose and plastic based membranes are compatible with a wide range of chemical agents and remain stable within a broad pH range. Used in a wide range of biological fields, the tangential flow cassettes guarantee high flow rates, low hold-up volumes, and low protein binding.


10  results were found

SearchResultCount:"10"

Sort Results

List View Easy View

Rate These Search Results

Supplier: Cytiva (Formerly Pall Lab)
Description: T-series centramate cassettes with Omegaâ„¢ membrane are ideal for development, pilot, and production scale TFF applications in diverse biological and b...

Supplier: Cytiva (Formerly Pall Lab)
Description: The disposable Minimateâ„¢ TFF capsules, with Omegaâ„¢ membrane, can perform concentration processes desalting or buffer exchange on the same system with ...

Supplier: Cytiva (Formerly Pall Lab)
Description: Tangential flow cassette, Centramate™, Membrane: Supor®, Filtration area: 200 cm², Pore size: 0,65 µm

Supplier: Cytiva (Formerly Pall Lab)
Description: Tangential flow cassette, Centramate™, Filtration area: 1000 cm², Pore size: 0,1 µm

Supplier: Sartorius
Description: Hydrosart® membranes are stabilised cellulose based membranes that have been optimised for the biotechnological and pharmaceutical industry. It is a s...

Catalog Number: (SPECXP300MP1L)
Supplier: Spectrum Laboratories
Description: SIUS PD cassette with HyStream membrane type and molecular weight cut-off (MWCO) of 300kD.
UOM: 1 * 1 items

New Product


Supplier: Sartorius
Description: Vivaflow® 50 and 200 devices are ready to use crossflow cassettes for filtration and concentration of samples from 100 to 3000 ml or 500 to 5000 ml. ...

Supplier: Sartorius
Description: Sartocon® Slice cassettes are made of the same materials and construction and, therefore, the identical flow path length as the Sartocon cassettes, us...

Supplier: Sartorius
Description: Polyethersulfone membrane
The robust polyethersulfone membrane (PESU) is a polymer, stable within a broad pH (1 to 14) and temperature ran...

Supplier: Sartorius
Description: Tangential flow cassette, Sartocon®, Membrane: Hydrosart®, Filtration area: 1000 cm², Pore size: 0,2 µm

Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on 0800 22 33 44.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on 0800 22 33 44
Additional Documentation may be needed to purchase this item. A VWR representative will contact you if needed.
Additional Documentation may be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
Product(s) marked with this symbol are discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalogue Number listed above. If you need further assistance, please call VWR Customer Service on 0800 22 33 44.
1 - 10 of 10
no targeter for Bottom