Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (1)
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Syringe Dispensers

https://uk.vwr.com/stibo/smallweb/std.lang.all/96/74/18709674.jpg

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"4"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  HAMILTON BONADUZ
Description:   The Microlab® 600 dispensers are highly precise syringe pumps with a touchscreen graphical user interface designed to quickly and easily dispense liqu...
Supplier:  HAMILTON BONADUZ
Description:   This multi-syringe dispenser consists of six identical 1700 RN series Gastight® syringes. An easy to use mechanism releases step by step movement of t...
Catalog Number: (549-0233)
Supplier:  HAMILTON BONADUZ
Description:   The PB600-1 works with Microliter™ or Gastight® syringes from 10 µl to 2,5 ml. Use the repeating dispenser with cemented needles (N), removable needle...
UOM:  1 * 1 items
Catalog Number: (738-0054)
Supplier:  GREINER BIO ONE
Description:   This micro-dispenser reduces the work required to dispense eosin and formalin solutions on 60-well micro-plates. It enables reagents to be added in si...
UOM:  1 * 100 items
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on 0800 22 33 44.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on 0800 22 33 44
Additional Documentation may be needed to purchase this item. A VWR representative will contact you if needed.
Additional Documentation may be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalogue Number listed above. If you need further assistance, please call VWR Customer Service on 0800 22 33 44.
1 - 4  of 4