Ice pans, rectangular
Supplier: Bel-Art
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
216-6930EA 226 GBP
BELAM188489101 BELAM188489102
Ice pans, rectangular
Coloured expanded polymer
  • Temperature range: –196 and +93 °C
  • Chemical resistant, impervious to moisture and odors, extremely robust
  • Durable, stackable, lightweight
  • Temperature range: –196 and +93 °C
  • Chemical resistant, impervious to moisture and odors, extremely robust
  • Durable, stackable, lightweight
Ideal for storing and transporting temperature sensitive samples in ice, alcohol slurries, water baths and more.