Would you like to log in again?
Sharp disposal containers for infectious waste
Supplier: VWR Collection
PP, luminous yellow. For collecting, storing and disposing of sharp, dangerous or infected waste products, such as cannulae, syringes, scalpel blades, etc.
  • Puncture-proof, autoclavable and suitable for incineration
  • Transparent, tightly sealing lid with cannula removal device
  • Tortuous path design restricts hand entry
  • Stackable
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Capacity Description W×D×H Pk VWR Catalog Number Quantity
3,8 l Medium 254×178×127 mm 24 119-0051 Unavailable
30,3 l Extra large 286×286×349 mm 10 119-0055 Unavailable
unavailable This product is discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalogue Number listed above. If you need further assistance, please call VWR Customer Service on 0800 22 33 44.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
WRITE A REVIEW
Need help?
  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.
VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.
Please confirm the industry you work in
Example: Pharmaceutical
Please confirm your role
Example: Clinical Trial Assistant
Please confirm your location
Example: New York, NY
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty